Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isophthalonitrile 98%

Supplier: AMBEED, INC
Warning

A310400-100G A310400-500G A310400-25G A310400-1000G
76802-748EA 15 USD InStock
76802-748 76802-750 77306-022 77668-372
Isophthalonitrile 98%
Isophthalonitrile
Precaución: for Research Use Only


Formula: C₈H₄N₂
MW: 128.13 g/mol
Boiling Pt: 288
MDL Number: MFCD00001795
CAS Number: 626-17-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty