Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Gly-Cl 95%

Supplier: AMBEED, INC
Danger

A317259-250MG A317259-5G
76802-846EA 19 USD InStock
76802-846 77601-430
Fmoc-Gly-Cl 95%
Fmoc-Gly-Cl
Precaución: for Research Use Only


Formula: C₁₇H₁₄NO₃Cl
MW: 315.7 g/mol
MDL Number: MFCD00235814
CAS Number: 103321-49-9
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty