Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloroethyl laurate 97%

Supplier: AMBEED, INC
Warning

A319341-100G A319341-1G A319341-25G A319341-5G
76802-868EA 187 USD InStock
76802-868 76802-870 76802-872 76802-874
2-Chloroethyl laurate 97%
2-Chloroethyl laurate
Precaución: for Research Use Only


Formula: C₁₄H₂₇ClO₂
MDL Number: MFCD00059475
CAS Number: 64919-15-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty