Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Pyridinecarboxylic acid hydrazide 4-amino-2-hydroxybenzoate 98%

Supplier: AMBEED, INC
Warning

A321390-100MG A321390-1MG
76802-908EA 10 USD InStock
76802-908 76802-910
4-Pyridinecarboxylic acid hydrazide 4-amino-2-hydroxybenzoate 98%
4-Pyridinecarboxylic acid hydrazide 4-amino-2-hydroxybenzoate
Precaución: for Research Use Only


Formula: C₁₃H₁₄N₄O₄
MDL Number: MFCD00079376
CAS Number: 2066-89-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty