Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

p-Tolyldiphenylphosphine 98%

Supplier: AMBEED, INC
Warning

A333408-10G A333408-25G
76803-072EA 24 USD InStock
76803-072 76803-074
p-Tolyldiphenylphosphine 98%
p-Tolyldiphenylphosphine
Precaución: for Research Use Only


Formula: C₁₉H₁₇P
MDL Number: MFCD00008543
CAS Number: 1031-93-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty