Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-furoate 96%

Supplier: AMBEED, INC
Danger

A338348-1G A338348-25G A338348-5G
76803-096EA 7 USD InStock
76803-096 76803-098 76803-100
Ethyl-3-furoate 96%
Ethyl-3-furoate
Precaución: for Research Use Only


Formula: C₇H₈O₃
MDL Number: MFCD00005348
CAS Number: 614-98-2
UN: 3272
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty