Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indole-3-lactic acid 97%

Supplier: AMBEED, INC
Warning

A349216-100MG A349216-1G A349216-250MG
76803-242EA 88 USD InStock
76803-242 76803-244 76803-246
Indole-3-lactic acid 97%
Indole-3-lactic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₃
MDL Number: MFCD00005642
CAS Number: 1821-52-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty