Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Di-tert-butyl-N,N-diethylphosphoramidite 98%

Supplier: AMBEED, INC
Warning

A350043-100G A350043-1G A350043-25G A350043-5G
76803-268EA 334 USD InStock
76803-268 76803-270 76803-272 76803-274
Di-tert-butyl-N,N-diethylphosphoramidite 98%
Di-tert-butyl-N,N-diethylphosphoramidite
Precaución: for Research Use Only


Formula: C₁₂H₂₈NO₂P
MW: 249.335 g/mol
MDL Number: MFCD00134307
CAS Number: 117924-33-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty