Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl palmitate 97%

Supplier: AMBEED, INC
Warning

A353232-100G A353232-1MG A353232-25G A353232-500G
76803-328EA 8 USD InStock
76803-328 76803-330 76803-332 76803-334
Ethyl palmitate 97%
Ethyl palmitate
Precaución: for Research Use Only


Formula: C₁₈H₃₆O₂
MDL Number: MFCD00008996
CAS Number: 628-97-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty