Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl allylphosphonate 98%

Supplier: AMBEED, INC
Warning

A355658-100G A355658-1G A355658-25G A355658-5G
76803-382EA 380 USD InStock
76803-382 76803-384 76803-386 76803-388
Diethyl allylphosphonate 98%
Diethyl allylphosphonate
Precaución: for Research Use Only


Formula: C₇H₁₅O₃P
MW: 178.1660 g/mol
MDL Number: MFCD00015134
CAS Number: 1067-87-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty