Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexaketocyclohexane octahydrate 99%

Supplier: AMBEED, INC
Warning

A373274-100G A373274-10G A373274-1G A373274-25G A373274-5G
76803-612EA 263 USD InStock
76803-612 76803-614 76803-616 76803-618 76803-620
Hexaketocyclohexane octahydrate 99%
Hexaketocyclohexane octahydrate
Precaución: for Research Use Only


Formula: C₆O₆·8H₂O
MW: 312.18 g/mol
MDL Number: MFCD00149074
CAS Number: 7255-28-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty