Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-m-anisate 98%

Supplier: AMBEED, INC
Warning

A373536-100G A373536-10G A373536-25G A373536-5G
76803-628EA 80 USD InStock
76803-628 76803-630 76803-632 76803-634
Ethyl-m-anisate 98%
Ethyl-m-anisate
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₃
MDL Number: MFCD00015149
CAS Number: 10259-22-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty