Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Amino-2-naphthoic acid 97%

Supplier: AMBEED, INC
Warning

A375169-100MG A375169-1G A375169-250MG A375169-25G A375169-5G
76803-656EA 5 USD InStock
76803-656 76803-658 76803-660 76803-662 76803-664
6-Amino-2-naphthoic acid 97%
6-Amino-2-naphthoic acid
Precaución: for Research Use Only


Formula: C₁₁H₉NO₂
MDL Number: MFCD01861831
CAS Number: 116668-47-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty