Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl (4-nitrobenzyl)phosphonate 95%

Supplier: AMBEED, INC
Warning

A375447-1G A375447-250MG A375447-5G
76803-670EA 18 USD InStock
76803-670 76803-672 76803-674
Diethyl (4-nitrobenzyl)phosphonate 95%
Diethyl (4-nitrobenzyl)phosphonate
Precaución: for Research Use Only


Formula: C₁₁H₁₆NO₅P
MW: 273.225 g/mol
MDL Number: MFCD00047747
CAS Number: 2609-49-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty