Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl benzylmalonate 98%

Supplier: AMBEED, INC
Warning

A390976-1000G A390976-100G A390976-10G A390976-25G A390976-500G A390976-5G
76803-858EA 194 USD InStock
76803-858 76803-860 76803-862 76803-864 76803-866 76803-868
Diethyl benzylmalonate 98%
Diethyl benzylmalonate
Precaución: for Research Use Only


Formula: C₁₄H₁₈O₄
MW: 250.29 g/mol
MDL Number: MFCD00009166
CAS Number: 607-81-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty