Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methoxy-4-nitronaphthalene 97%

Supplier: AMBEED, INC
Warning

A393374-10G A393374-1G A393374-25G A393374-5G
76803-878EA 59 USD InStock
76803-878 76803-880 76803-882 76803-884
1-Methoxy-4-nitronaphthalene 97%
1-Methoxy-4-nitronaphthalene
Precaución: for Research Use Only


Formula: C₁₁H₉NO₃
MDL Number: MFCD00003926
CAS Number: 4900-63-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty