Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl acetylenedicarboxylate 98% (by GC)

Supplier: AMBEED, INC
Danger

A401572-100G A401572-10G A401572-25G
76803-926EA 43 USD InStock
76803-926 76803-928 76803-930
Dimethyl acetylenedicarboxylate 98% (by GC)
Dimethyl acetylenedicarboxylate
Precaución: for Research Use Only


Formula: C₆H₆O₄
MW: 142.11 g/mol
MDL Number: MFCD00008456
CAS Number: 762-42-5
UN: 1760
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty