Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isobutyl hexanoate 98%

Supplier: AMBEED, INC
Warning

A404422-100G A404422-25G A404422-500G
76803-950EA 17 USD InStock
76803-950 76803-952 76803-954
Isobutyl hexanoate 98%
Isobutyl hexanoate
Precaución: for Research Use Only


Formula: C₁₀H₂₀O₂
MDL Number: MFCD00048870
CAS Number: 105-79-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty