Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydroquinine 97%

Supplier: AMBEED, INC
Warning

A407037-100G A407037-100MG A407037-10G A407037-1G A407037-250MG A407037-25G A407037-5G
76803-986EA 427 USD InStock
76803-986 76803-988 76803-990 76803-992 76803-994 76803-996 76803-998
Hydroquinine 97%
Hydroquinine
Precaución: for Research Use Only


Formula: C₂₀H₂₆N₂O₂
MDL Number: MFCD00151107
CAS Number: 522-66-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty