Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethoxyacetonitrile 97%

Supplier: AMBEED, INC
Danger

A410422-10G A410422-1G A410422-25G A410422-5G
76804-036EA 15 USD InStock
76804-036 76804-038 76804-040 76804-042
Diethoxyacetonitrile 97%
Diethoxyacetonitrile
Precaución: for Research Use Only


Formula: C₆H₁₁NO₂
MDL Number: MFCD00043982
CAS Number: 6136-93-2
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty