Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl difluoromalonate 98%

Supplier: AMBEED, INC
Danger

A427563-1G A427563-25G A427563-5G
76804-242EA 18 USD InStock
76804-242 76804-244 76804-246
Diethyl difluoromalonate 98%
Diethyl difluoromalonate
Precaución: for Research Use Only


Formula: C₇H₁₀F₂O₄
MW: 196.15 g/mol
MDL Number: MFCD00221617
CAS Number: 680-65-9
UN: 2810
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty