Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium laurate 98%

Supplier: AMBEED, INC
Danger

A428676-25G A428676-100G
76804-264EA 7 USD InStock
76804-264 77601-850
Sodium laurate 98%
Sodium laurate
Precaución: for Research Use Only


Formula: C₁₂H₂₃NaO₂
MW: 222.304 g/mol
MDL Number: MFCD00041754
CAS Number: 629-25-4
UN: 1759
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty