Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-2-aminobutyric acid 95%

Supplier: AMBEED, INC
Warning

A429744-10G A429744-250MG A429744-25G A429744-5G
76804-266EA 65 USD InStock
76804-266 76804-268 76804-270 76804-272
Fmoc-2-aminobutyric acid 95%
Fmoc-2-aminobutyric acid
Precaución: for Research Use Only


Formula: C₁₉H₁₉NO₄
MDL Number: MFCD00270362
CAS Number: 174879-28-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty