Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-6-methylcoumarin 98%

Supplier: AMBEED, INC
Warning

A431117-1G A431117-250MG A431117-5G
76804-282EA 30 USD InStock
76804-282 76804-284 76804-286
4-Hydroxy-6-methylcoumarin 98%
4-Hydroxy-6-methylcoumarin
Precaución: for Research Use Only


Formula: C₁₀H₈O₃
MDL Number: MFCD00085738
CAS Number: 13252-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty