Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,2R)-3,3-Disulfanediylbis(2-aminopropanoic acid) xhydrochloride 98%

Supplier: AMBEED, INC
Danger

A433879-10G A433879-25G
76804-316EA 14 USD InStock
76804-316 76804-318
(2R,2R)-3,3-Disulfanediylbis(2-aminopropanoic acid) xhydrochloride 98%
(2R,2R)-3,3-Disulfanediylbis(2-aminopropanoic acid) xhydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₃ClN₂O₄S₂
MDL Number: MFCD00068399
CAS Number: 34760-60-6
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty