Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-1-naphthoic acid 95%

Supplier: AMBEED, INC
Warning

A444820-100MG A444820-1G A444820-250MG A444820-5G
76804-420EA 8 USD InStock
76804-420 76804-422 76804-424 76804-426
2-Methyl-1-naphthoic acid 95%
2-Methyl-1-naphthoic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₀O₂
MW: 186.206 g/mol
MDL Number: MFCD00089618
CAS Number: 1575-96-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty