Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl diethyldithiocarbamate 95%

Supplier: AMBEED, INC
Warning

A446461-100MG A446461-1G A446461-250MG A446461-5G
76804-456EA 29 USD InStock
76804-456 76804-458 76804-460 76804-462
Benzyl diethyldithiocarbamate 95%
Benzyl diethyldithiocarbamate
Precaución: for Research Use Only


Formula: C₁₂H₁₇NS₂
MDL Number: MFCD00191425
CAS Number: 3052-61-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty