Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Heptaethylene glycol 96%

Supplier: AMBEED, INC
Warning

A449474-100MG A449474-10G A449474-1G A449474-250MG A449474-5G A449474-100G
76804-506EA 9 USD InStock
76804-506 76804-508 76804-510 76804-512 76804-514 77669-020
Heptaethylene glycol 96%
Heptaethylene glycol
Precaución: for Research Use Only


Formula: C₁₄H₃₀O₈
MDL Number: MFCD00002876
CAS Number: 5617-32-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty