Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1-hydroxy-1H-1,2,3-triazole-4-carboxylate 98%

Supplier: AMBEED, INC
Danger

A460265-1G A460265-25G A460265-5G
76804-628EA 32 USD InStock
76804-628 76804-630 76804-632
Ethyl-1-hydroxy-1H-1,2,3-triazole-4-carboxylate 98%
Ethyl-1-hydroxy-1H-1,2,3-triazole-4-carboxylate
Precaución: for Research Use Only


Formula: C₅H₇N₃O₃
MDL Number: MFCD06796746
CAS Number: 137156-41-3
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty