Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl diphenylacetate 95%

Supplier: AMBEED, INC
Warning

A468933-1G A468933-250MG A468933-25G A468933-5G
76804-734EA 45 USD InStock
76804-734 76804-736 76804-738 76804-740
Ethyl diphenylacetate 95%
Ethyl diphenylacetate
Precaución: for Research Use Only


Formula: C₁₆H₁₆O₂
MDL Number: MFCD00026861
CAS Number: 3468-99-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty