Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methylbenzofuran-2-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A471801-100G A471801-10G A471801-1G A471801-25G A471801-5G
76804-746EA 171 USD InStock
76804-746 76804-748 76804-750 76804-752 76804-754
3-Methylbenzofuran-2-carboxylic acid 98%
3-Methylbenzofuran-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₈O₃
MW: 176.17 g/mol
MDL Number: MFCD00060511
CAS Number: 24673-56-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty