Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7,8-Dimethylalloxazine 95%

Supplier: AMBEED, INC
Warning

A484292-100MG A484292-1G A484292-250MG A484292-5G
76804-842EA 27 USD InStock
76804-842 76804-844 76804-846 76804-848
7,8-Dimethylalloxazine 95%
7,8-Dimethylalloxazine
Precaución: for Research Use Only


Formula: C₁₂H₁₀N₄O₂
MDL Number: MFCD00005021
CAS Number: 1086-80-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty