Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl alfa-amino butyrate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A484831-1G A484831-25G A484831-5G
76804-850EA 21 USD InStock
76804-850 76804-852 76804-854
Ethyl alfa-amino butyrate hydrochloride 98%
Ethyl alfa-amino butyrate hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₃NO₂·ClH
MW: 167.634 g/mol
MDL Number: MFCD08689937
CAS Number: 55410-21-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty