Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dihexadecyl hydrogen phosphate 97%

Supplier: AMBEED, INC
Warning

A492844-100MG A492844-1G A492844-250MG A492844-5G
76804-904EA 22 USD InStock
76804-904 76804-906 76804-908 76804-910
Dihexadecyl hydrogen phosphate 97%
Dihexadecyl hydrogen phosphate
Precaución: for Research Use Only


Formula: C₃₂H₆₇O₄P
MDL Number: MFCD00008999
CAS Number: 2197-63-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty