Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-butylacetoacetate 95%

Supplier: AMBEED, INC
Warning

A493755-100G A493755-1G A493755-25G A493755-5G
76804-924EA 92 USD InStock
76804-924 76804-926 76804-928 76804-930
Ethyl-2-butylacetoacetate 95%
Ethyl-2-butylacetoacetate
Precaución: for Research Use Only


Formula: C₁₀H₁₈O₃
MW: 186.25 g/mol
MDL Number: MFCD00026888
CAS Number: 1540-29-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty