Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-3,4,5-trifluoro-L-phenylalanine 98%

Supplier: AMBEED, INC
Warning

A499178-1G A499178-250MG
76804-962EA 234 USD InStock
76804-962 77669-244
Fmoc-3,4,5-trifluoro-L-phenylalanine 98%
Fmoc-3,4,5-trifluoro-L-phenylalanine
Precaución: for Research Use Only


Formula: C₂₄H₁₈F₃NO₄
MW: 441.41 g/mol
MDL Number: MFCD00797583
CAS Number: 205526-30-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty