Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium (S)-5-oxopyrrolidine-2-carboxylate 97%

Supplier: AMBEED, INC
Warning

A502870-100G A502870-25G A502870-500G
76805-004EA 23 USD InStock
76805-004 76805-006 76805-008
Sodium (S)-5-oxopyrrolidine-2-carboxylate 97%
Sodium (S)-5-oxopyrrolidine-2-carboxylate
Precaución: for Research Use Only


Formula: C₅H₆NNaO₃
MW: 151.09 g/mol
MDL Number: MFCD01749005
CAS Number: 28874-51-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty