Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium tetrachloroaurate 98%

Supplier: AMBEED, INC
Danger

A505661-1G A505661-5G
76805-010EA 138 USD InStock
76805-010 76805-012
Sodium tetrachloroaurate 98%
Sodium tetrachloroaurate
Precaución: for Research Use Only


Formula: AuCl₄Na
MW: 361.77
MDL Number: MFCD00003473
CAS Number: 15189-51-2
UN: 2811
ADR: 6.1 Ⅲ
Merck Index: 12,08837
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty