Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Nitroquinaldine 98%

Supplier: AMBEED, INC
Warning

A508072-100MG A508072-1G A508072-25G
76805-030EA 9 USD InStock
76805-030 76805-032 76805-034
6-Nitroquinaldine 98%
6-Nitroquinaldine
Precaución: for Research Use Only


Formula: C₁₀H₈N₂O₂
MDL Number: MFCD00051736
CAS Number: 613-30-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty