Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glycocyamine 95%

Supplier: AMBEED, INC
Warning

A515873-100G A515873-25G A515873-500G A515873-1000G
76805-120EA 13 USD InStock
76805-120 76805-122 76805-124 77669-292
Glycocyamine 95%
Glycocyamine
Precaución: for Research Use Only


Formula: C₃H₇N₃O₂
MDL Number: MFCD00004278
CAS Number: 352-97-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty