Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Acetoxyindole 95%

Supplier: AMBEED, INC
Warning

A516757-1G A516757-250MG A516757-25G A516757-5G
76805-128EA 25 USD InStock
76805-128 76805-130 76805-132 76805-134
3-Acetoxyindole 95%
3-Acetoxyindole
Precaución: for Research Use Only


Formula: C₁₀H₉NO₂
MDL Number: MFCD00014561
CAS Number: 608-08-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty