Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dibenzyloxybenzoic acid 96%

Supplier: AMBEED, INC
Warning

A518874-100G A518874-10G A518874-1G A518874-25G A518874-500G A518874-5G
76805-140EA 88 USD InStock
76805-140 76805-142 76805-144 76805-146 76805-148 76805-150
3,5-Dibenzyloxybenzoic acid 96%
3,5-Dibenzyloxybenzoic acid
Precaución: for Research Use Only


Formula: C₂₁H₁₈O₄
MW: 334.37 g/mol
MDL Number: MFCD00191341
CAS Number: 28917-43-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty