Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl trifluoromethanesulphonate 99%

Supplier: AMBEED, INC
Danger

A525593-25G A525593-5G
76805-214EA 71 USD InStock
76805-214 76805-216
Ethyl trifluoromethanesulphonate 99%
Ethyl trifluoromethanesulphonate
Precaución: for Research Use Only


Formula: C₃H₅F₃O₃S
MDL Number: MFCD00000410
CAS Number: 425-75-2
UN: 2920
ADR: 8,3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty