Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl pimelate 98%

Supplier: AMBEED, INC
Warning

A531226-100G A531226-25G A531226-5G
76805-278EA 89 USD InStock
76805-278 76805-280 76805-282
Diethyl pimelate 98%
Diethyl pimelate
Precaución: for Research Use Only


Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
Boiling Pt: 268
Melting Pt: -24
MDL Number: MFCD00009216
CAS Number: 2050-20-6
Merck Index: 12,07585
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty