Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Carbobenzoxy-DL-serine 97%

Supplier: AMBEED, INC
Warning

A535096-100G A535096-10G A535096-1G A535096-25G
76805-322EA 168 USD InStock
76805-322 76805-324 76805-326 76805-328
N-Carbobenzoxy-DL-serine 97%
N-Carbobenzoxy-DL-serine
Precaución: for Research Use Only


Formula: C₁₁H₁₃NO₅
MW: 239.23 g/mol
MDL Number: MFCD00063143
CAS Number: 2768-56-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty