Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Manganese gluconate 95%

Supplier: AMBEED, INC
Warning

A537304-100G A537304-500G A537304-5G
76805-336EA 40 USD InStock
76805-336 76805-338 77602-290
Manganese gluconate 95%
Manganese gluconate
Precaución: for Research Use Only


Formula: C₁₂H₂₂MnO₁₄
MW: 445.2327 g/mol
MDL Number: MFCD00867656
CAS Number: 6485-39-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty