Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Disodium fluorophosphate 95%

Supplier: AMBEED, INC
Warning

A537600-1000G A537600-100G A537600-500G
76805-340EA 20 USD InStock
76805-340 76805-342 76805-344
Disodium fluorophosphate 95%
Disodium fluorophosphate
Precaución: for Research Use Only


Formula: FNa₂O₃P
MW: 143.95 g/mol
MDL Number: MFCD00014248
CAS Number: 10163-15-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty