Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Gemifloxacin mesylate 98%

Supplier: AMBEED, INC
Warning

A538822-100MG A538822-10MG A538822-1MG A538822-25MG A538822-50MG
76805-350EA 213 USD InStock
76805-350 76805-352 76805-354 76805-356 76805-358
Gemifloxacin mesylate 98%
Gemifloxacin mesylate
Precaución: for Research Use Only


Formula: C₁₈H₂₀FN₅O₄·CH₄O₃S
MW: 485.49 g/mol
MDL Number: MFCD08277637
CAS Number: 210353-53-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty