Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-cyanobutanoate 98%

Supplier: AMBEED, INC
Warning

A542893-1G A542893-250MG A542893-5G
76805-392EA 70.02 USD InStock
76805-392 76805-394 76805-396 76805-398
Ethyl-2-cyanobutanoate 98%
Ethyl-2-cyanobutanoate
Precaución: for Research Use Only


Formula: C₇H₁₁NO₂
MDL Number: MFCD02258598
CAS Number: 1619-58-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty