Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl bicyclo[2.2.1]hept-5-ene-2-carboxylate 98%

Supplier: AMBEED, INC
Warning

A548406-100G A548406-25G A548406-500G A548406-5G
76805-438EA 245 USD InStock
76805-438 76805-440 76805-442 76805-444
Ethyl bicyclo[2.2.1]hept-5-ene-2-carboxylate 98%
Ethyl bicyclo[2.2.1]hept-5-ene-2-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₄O₂
MW: 166.21 g/mol
MDL Number: MFCD00167594
CAS Number: 10138-32-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty